Methyl (4-piperidyl)acetate hydrochloride ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: (4-Piperidyl)acetic Acid Methyl Ester Hydrochloride, Methyl 2-(piperidin-4-yl)acetate hydrochloride

M1926-1G M1926-5G
TCIAM1926-1GEA 0 CNY
TCIAM1926-1G TCIAM1926-5G
Methyl (4-piperidyl)acetate hydrochloride ≥98.0% (by titrimetric analysis)
Methyl (4-piperidyl)acetate hydrochloride
分子式: C₈H₁₅NO₂
分子量: 193.67 g/mol
MDL编号: MFCD09261419
CAS号: 81270-37-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 153.0 to 157.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR