Methyl-3-chloro-2,2,3,3-tetrafluoropropionate ≥97.0% (by GC)

供应商: TCI
Danger

Synonyms: 3-Chloro-2,2,3,3-tetrafluoropropionic Acid Methyl Ester

M1902-1G M1902-5G
TCIAM1902-1GEA 0 CNY
TCIAM1902-1G TCIAM1902-5G
Methyl-3-chloro-2,2,3,3-tetrafluoropropionate ≥97.0% (by GC)
Methyl-3-chloro-2,2,3,3-tetrafluoropropionate
分子式: C₄H₃ClF₄O₂
分子量: 194.51 g/mol
MDL编号: MFCD00155715
CAS号: 127589-63-3
UN号: 2924
ADR: 3 II

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR