Methyl-1,2,3,4-tetra-O-acetyl-β-D-glucuronate ≥96.0% (by GC)

供应商: TCI

Synonyms: Methyl 1,2,3,4-Tetra-O-acetyl-β-D-glucopyranuronate, (2S,3R,4S,5S,6S)-6-(Methoxycarbonyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate, 1,2,3,4-Tetra-O-acetyl-β-D-glucuronic acid Methyl Ester, 1,2,3,4-Tetra-O-acetyl-β-D-glucopyranuronic acid Methyl Ester, Methyl tetra-O-acetyl-β-D-glucopyranuronate

M1868-1G M1868-5G
TCIAM1868-1GEA 0 CNY
TCIAM1868-1G TCIAM1868-5G
Methyl-1,2,3,4-tetra-O-acetyl-β-D-glucuronate ≥96.0% (by GC)
Methyl-1,2,3,4-tetra-O-acetyl-beta-D-glucuronate
分子式: C₁₅H₂₀O₁₁
分子量: 376.31 g/mol
MDL编号: MFCD00069834
CAS号: 7355-18-2
EINECS: 230-880-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 177.0 to 181.0 deg-C
Specific rotation [a]20/D +7.0 ~ +9.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR