Methyl-2-oxovalerate ≥90.0% (通过GC)
供应商: TCI
M1863-25G
M1863-5G
TCIAM1863-25GEA
0
CNY
TCIAM1863-25G
TCIAM1863-5G
Methyl-2-oxovalerate ≥90.0% (通过GC)
Methyl-2-oxovalerate
分子式:
C₆H₁₀O₃ 沸点: 62°C 存储条件: Refrigerator |
MDL编号:
MFCD10566912 CAS号: 6376-59-6 |
产品技术规格
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...