4-Methyl-4-nitrovaleraldehyde ≥90.0% (通过GC)

供应商: TCI
Warning

M1825-25G M1825-5G
TCIAM1825-25GEA 0 CNY
TCIAM1825-25G TCIAM1825-5G
4-Methyl-4-nitrovaleraldehyde ≥90.0% (通过GC)
4-Methyl-4-nitrovaleraldehyde
分子式: C₆H₁₁NO₃
沸点: 90°C
CAS号: 57620-49-2

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR