Methyl Indole-2-carboxylate ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: Methyl 1H-indole-2-carboxylate, Indole-2-carboxylic Acid Methyl Ester

M1749-25G M1749-5G
TCIAM1749-25GEA 0 CNY
TCIAM1749-25G TCIAM1749-5G
Methyl Indole-2-carboxylate ≥98.0% (by GC)
Methyl Indole-2-carboxylate
分子式: C₁₀H₉NO₂
分子量: 175.19 g/mol
MDL编号: MFCD00460779
CAS号: 1202-04-6
EINECS: 214-861-7

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR