Methyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥98.0% (by GC)
供应商: TCI
M1682-1G
M1682-5G
TCIAM1682-1GEA
0
CNY
TCIAM1682-1G
TCIAM1682-5G
Methyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥98.0% (by GC)
Methyl-2,3,4,6-tetra-O-acetyl-1-thio-beta-D-glucopyranoside
分子式:
C₁₅H₂₂O₉S 分子量: 378.39 g/mol 存储条件: Refrigerator |
CAS号:
13350-45-3 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -13.0 to -17.0 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...