2-Methoxy-1-butanol ≥98.0% (by GC)
供应商: TCI
|
Warning
|
M1497-25G
M1497-5G
TCIAM1497-25GEA
0
CNY
TCIAM1497-25G
TCIAM1497-5G
2-Methoxy-1-butanol ≥98.0% (by GC)
2-Methoxy-1-butanol
分子式:
C₅H₁₂O₂ 沸点: 146°C |
CAS号:
15467-25-1 EINECS: 239-486-6 UN号: 1993 ADR: 3,III |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...