3-甲氨基-1-丙醇 ≥97.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
3-(Methylamino)propan-1-ol
M1484-25G
M1484-5G
TCIAM1484-25GEA
0
CNY
TCIAM1484-25G
TCIAM1484-5G
3-甲氨基-1-丙醇 ≥97.0% (by GC, titration analysis)
3-甲氨基-1-丙醇
分子式:
C₄H₁₁NO 沸点: 169°C |
MDL编号:
MFCD06797456 CAS号: 42055-15-2 UN号: 2735 ADR: 8,III |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...