6-Methoxy-4-methylcoumarin ≥98.0% (by GC)
供应商: TCI
M1398-25G
M1398-5G
TCIAM1398-25GEA
0
CNY
TCIAM1398-25G
TCIAM1398-5G
6-Methoxy-4-methylcoumarin ≥98.0% (by GC)
6-Methoxy-4-methylcoumarin
分子式:
C₁₁H₁₀O₃ |
MDL编号:
MFCD00635101 CAS号: 6295-35-8 |
产品技术规格
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 163.0 to 166.0 deg-C |
Solubility in hot DMF | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...