N-Methyl-4-nitrophthalimide ≥99.0% (by HPLC)

供应商: TCI

Synonyms: 2-Methyl-5-nitroisoindoline-1,3-dione

M1310-25G M1310-5G
TCIAM1310-25GEA 0 CNY
TCIAM1310-25G TCIAM1310-5G
N-Methyl-4-nitrophthalimide ≥99.0% (by HPLC)
N-Methyl-4-nitrophthalimide
分子式: C₉H₆N₂O₄
分子量: 206.16 g/mol
MDL编号: MFCD00454263
CAS号: 41663-84-7
EINECS: 255-483-2

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 99.0 area%
Melting point 177.0 to 180.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR