2,2'-Methylenebis[6-(1-methylcyclohexyl)-p-cresol] ≥95.0% (通过滴定分析)

供应商: TCI
Warning

M1301-25G M1301-500G
TCIAM1301-25GEA 0 CNY
TCIAM1301-25G TCIAM1301-500G
2,2'-Methylenebis[6-(1-methylcyclohexyl)-p-cresol] ≥95.0% (通过滴定分析)
2,2'-Methylenebis[6-(1-methylcyclohexyl)-p-cresol]
分子式: C₂₉H₄₀O₂
MDL编号: MFCD00151797
CAS号: 77-62-3
EINECS: 201-044-5

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 90.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 136.0 to 140.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR