(甲氧基羰基氨磺酰基)三乙基氢氧化铵, ≥97.0% (by total nitrogen basis)

供应商: TCI
Warning

Synonyms: N-(三乙基氨磺酰)氨基甲酸甲酯, 伯吉斯试剂

M1279-1G M1279-25G M1279-5G
TCIAM1279-1GEA 0 CNY
TCIAM1279-1G TCIAM1279-25G TCIAM1279-5G
(甲氧基羰基氨磺酰基)三乙基氢氧化铵, ≥97.0% (by total nitrogen basis)
(甲氧基羰基氨磺酰基)三乙基氢氧化铵,
分子式: C₈H₁₈N₂O₄S
分子量: 238.30 g/mol
存储条件: Refrigerator
MDL编号: MFCD00077815
CAS号: 29684-56-8
Merck Index: 14,01502

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Melting point 74.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR