N-Methyl-1-naphthylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 1-(N-Methylamino)naphthalene Hydrochloride

M1172-25G M1172-5G
TCIAM1172-25GEA 0 CNY
TCIAM1172-25G TCIAM1172-5G
N-Methyl-1-naphthylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
N-Methyl-1-naphthylamine hydrochloride
分子式: C₁₁H₁₁N
分子量: 193.67 g/mol
MDL编号: MFCD00191555
CAS号: 4643-36-1

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR