Dibutyl methylenebis(thioglycolate) ≥95.0% (by GC)

供应商: TCI

Synonyms: Methylenebis(thioglycolic Acid Dibutyl Ester)

M1137-25G M1137-500G
TCIAM1137-25GEA 0 CNY
TCIAM1137-25G TCIAM1137-500G
Dibutyl methylenebis(thioglycolate) ≥95.0% (by GC)
Dibutyl methylenebis(thioglycolate)
分子式: C₁₃H₂₄O₄S₂
分子量: 308.45 g/mol
沸点: 178°C
闪点: 167°C
MDL编号: MFCD00027216
CAS号: 14338-82-0
EINECS: 238-289-2

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR