(+)-(4,6-O-亚苄基)甲基-α-D-吡喃葡萄糖苷 ≥98.0% (by GC)

供应商: TCI

Synonyms: 4,6-O-亚苄基-甲基-α-D-吡喃葡萄糖苷, 甲基 4,6-O-亚苄基-α-D-吡喃葡萄糖苷

M1125-25G M1125-5G
TCIAM1125-25GEA 0 CNY
TCIAM1125-25G TCIAM1125-5G
(+)-(4,6-O-亚苄基)甲基-α-D-吡喃葡萄糖苷 ≥98.0% (by GC)
(+)-(4,6-O-亚苄基)甲基-α-D-吡喃葡萄糖苷
分子式: C₁₄H₁₈O₆
MDL编号: MFCD00006819
CAS号: 3162-96-7
EINECS: 221-615-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +100.0 to +110.0 deg (C=2, CH3CN)
Drying loss max. 4.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR