Ethyl 2-Methoxy-6-methylbenzoate ≥95.0% (by GC)

供应商: TCI

Synonyms: Ethyl 2-methoxy-6-methylbenzoate, 2-Methoxy-6-methylbenzoic Acid Ethyl Ester

M1120-25G M1120-5G
TCIAM1120-25GEA 0 CNY
TCIAM1120-25G TCIAM1120-5G
Ethyl 2-Methoxy-6-methylbenzoate ≥95.0% (by GC)
Ethyl-2-methoxy-6-methylbenzoate
分子式: C₁₁H₁₄O₃
分子量: 194.23 g/mol
沸点: 135°C
MDL编号: MFCD00191666
CAS号: 6520-83-8

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR