N-甲基-2-吡咯甲醛 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
N-Methylpyrrole-2-carboxaldehyde, 1-Methylpyrrole-2-aldehyde, 1-Methylpyrrole-2-carboxaldehyde, 1-Methylpyrrol-2-carbaldehyde, 2-Formyl-1-methylpyrrole
M1119-25G
M1119-5G
TCIAM1119-25GEA
0
CNY
TCIAM1119-25G
TCIAM1119-5G
N-甲基-2-吡咯甲醛 ≥97.0% (by GC)
N-甲基-2-吡咯甲醛
分子式:
C₆H₇NO 分子量: 109.13 g/mol 沸点: 90°C 闪点: 72°C |
MDL编号:
MFCD00003087 CAS号: 1192-58-1 EINECS: 214-755-0 |
产品技术规格
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...