4,4'-Methylenebis(2,6-dimethylphenol) ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: Bis(4-hydroxy-3,5-dimethylphenyl)methane, 4,4'-Dihydroxy-3,3',5,5'-tetramethyldiphenylmethane, 2,2',6,6'-Tetramethyl-4,4'-methylenediphenol

M1099-25G M1099-500G
TCIAM1099-25GEA 0 CNY
TCIAM1099-25G TCIAM1099-500G
4,4'-Methylenebis(2,6-dimethylphenol) ≥98.0% (by GC)
4,4'-Methylenebis(2,6-dimethylphenol)
分子式: C₁₇H₂₀O₂
分子量: 256.35 g/mol
MDL编号: MFCD00055640
CAS号: 5384-21-4
EINECS: 226-378-9

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 175.0 to 179.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR