4,4′-(1-苯乙基)双酚 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 双酚 AP

M1098-25G M1098-500G
TCIAM1098-25GEA 0 CNY
TCIAM1098-25G TCIAM1098-500G
4,4′-(1-苯乙基)双酚 ≥98.0% (by GC)
4,4′-(1-苯乙基)双酚
分子式: C₂₀H₁₈O₂
分子量: 290.36 g/mol
MDL编号: MFCD00134687
CAS号: 1571-75-1
EINECS: 433-130-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 186.0 to 190.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR