Methyl Jasmonate (mixture of isomers) ≥90.0% (by GC)

供应商: TCI

Synonyms: 2-(2-Pentenyl)-3-methoxycarbonylmethylcyclopentanone, Methyl (Z)-2-(3-oxo-2-(pent-2-en-1-yl)cyclopentyl)acetate, 3-Oxo-2-(2-pentenyl)cyclopentaneacetic Acid Methyl Ester, Jasmonic Acid Methyl Ester, Methyl 3-Oxo-2-(2-pentenyl)cyclopentaneacetate

M1068-25G M1068-5G
TCIAM1068-25GEA 0 CNY
TCIAM1068-25G TCIAM1068-5G
Methyl Jasmonate (mixture of isomers) ≥90.0% (by GC)
Methyl Jasmonate (mixture of isomers)
分子式: C₁₃H₂₀O₃
分子量: 224.30 g/mol
沸点: 96°C
闪点: 156°C
CAS号: 1101843-02-0
EINECS: 254-705-5

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR