1-(3-Methoxyphenoxy)-2-propanol ≥90.0% (by GC)
供应商: TCI
Synonyms:
3-(2-Hydroxypropoxy)anisole, 1-(3-Methoxyphenoxy)propan-2-ol
M1061-25G
M1061-5G
TCIAM1061-25GEA
0
CNY
TCIAM1061-25G
TCIAM1061-5G
1-(3-Methoxyphenoxy)-2-propanol ≥90.0% (by GC)
1-(3-Methoxyphenoxy)-2-propanol
分子式:
C₁₀H₁₄O₃ 沸点: 110°C |
MDL编号:
MFCD00191540 CAS号: 382141-68-6 |
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...