β-Methyllevulinic acid ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 3-Acetylbutyric Acid, 3-Methyl-4-oxopentanoic Acid

M1057-25G M1057-5G
TCIAM1057-25GEA 0 CNY
TCIAM1057-25G TCIAM1057-5G
β-Methyllevulinic acid ≥98.0% (by GC, titration analysis)
beta-Methyllevulinic acid
分子式: C₆H₁₀O₃
分子量: 130.14 g/mol
沸点: 242°C
存储条件: Refrigerator
MDL编号: MFCD00143691
CAS号: 6628-79-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 32.0 to 35.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR