Methyl-ɑ-D-galactopyranoside monohydrate ≥98.0% (by GC)

供应商: TCI

Synonyms: Methyl-ɑ-D-galactoside monohydrate

M1047-25G M1047-5G
TCIAM1047-25GEA 0 CNY
TCIAM1047-25G TCIAM1047-5G
Methyl-ɑ-D-galactopyranoside monohydrate ≥98.0% (by GC)
Methyl-alpha-D-galactopyranoside monohydrate
分子式: C₇H₁₄O₆·H₂O
分子量: 194.18 g/mol (anhy)
MDL编号: MFCD00064085
CAS号: 34004-14-3
EINECS: 222-251-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 106.0 to 110.0 deg-C
Specific rotation [a]20/D +173 to +183 deg (C=1, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR