Methoxybenzoquinone ≥99.0% (by GC)

供应商: TCI

Synonyms: 2-Methoxycyclohexa-2,5-diene-1,4-dione

M1045-1G M1045-5G
TCIAM1045-1GEA 0 CNY
TCIAM1045-1G TCIAM1045-5G
Methoxybenzoquinone ≥99.0% (by GC)
Methoxybenzoquinone
分子式: C₇H₆O₃
MDL编号: MFCD00144755
CAS号: 2880-58-2

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Purity (Iodometric Titration) min. 95.0 %
Melting point 145.0 to 149.0 deg-C
Solubility in hot Methanol within very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR