Methyl-β-D-galactopyranoside 98,0 GC_ASSAY_METHOD

供应商: TCI

Synonyms: Methyl-β-D-galactoside, (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol, (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

M1035-1G M1035-5G
TCIAM1035-1GEA 0 CNY
TCIAM1035-1G TCIAM1035-5G
Methyl-β-D-galactopyranoside 98,0 GC_ASSAY_METHOD
Methyl-beta-D-galactopyranoside
分子式: C₇H₁₄O₆
分子量: 194.18 g/mol
MDL编号: MFCD00064357
CAS号: 1824-94-8
EINECS: 217-361-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 175.0 to 179.0 deg-C
Specific rotation [a]20/D -15.0 to -18.0 deg (C=1.5, MeOH)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR