3-Methyl-3-heptene (cis- and trans- mixture) cis- and trans-mixture ≥97.0% (通过GC)
供应商: TCI
|
Danger
|
M0991-1ML
M0991-5ML
TCIAM0991-1MLEA
0
CNY
TCIAM0991-1ML
TCIAM0991-5ML
3-Methyl-3-heptene (cis- and trans- mixture) cis- and trans-mixture ≥97.0% (通过GC)
3-Methyl-3-heptene (cis- and trans- mixture)
分子式:
C₈H₁₆ 沸点: 121°C |
MDL编号:
MFCD00048796 CAS号: 7300-03-0 UN号: 3295 ADR: 3,II |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...