Methylcyclohexene-1,2-dicarboxylic anhydride (mixture of isomers) ≥80.0% (by GC)

供应商: TCI
Danger

Synonyms: Methyltetrahydrophthalic Anhydride

M0683-25G M0683-500G
TCIAM0683-25GEA 0 CNY
TCIAM0683-25G TCIAM0683-500G
Methylcyclohexene-1,2-dicarboxylic anhydride (mixture of isomers) ≥80.0% (by GC)
Methylcyclohexene-1,2-dicarboxylic anhydride (mixture of isomers)
分子式: C₉H₁₀O₃
分子量: 166.18 g/mol
闪点: 157°C
MDL编号: MFCD00014585
CAS号: 11070-44-3
EINECS: 234-290-7
UN号: 3082
ADR: 9 III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 80.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR