(甲磺酰)乙腈 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: Methanesulphonylacetonitrile, Cyanomethylsulphonylmethane, 2-(Methylsulphonyl)acetonitrile

M0667-25G M0667-5G
TCIAM0667-25GEA 0 CNY
TCIAM0667-25G TCIAM0667-5G
(甲磺酰)乙腈 ≥98.0% (by GC)
(甲磺酰)乙腈
分子式: C₃H₅NO₂S
分子量: 119.14 g/mol
MDL编号: MFCD00014743
CAS号: 2274-42-2
EINECS: 218-888-5
UN号: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c06cd7cd9cd4722327b592d201572c91782c30c3&vid=c06cd7cd9cd4722327b592d201572c91782c30c3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Green to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR