Calcium methanesulphonate ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: Methanesulphonic acid calcium salt

M0549-25G M0549-500G
TCIAM0549-25GEA 0 CNY
TCIAM0549-25G TCIAM0549-500G
Calcium methanesulphonate ≥98.0% (by titrimetric analysis)
Calcium methanesulphonate
分子式: CH₄O₃S
分子量: 230.26 g/mol
MDL编号: MFCD00070543
CAS号: 58131-47-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR