甲基丙烯酸正己酯 ≥98.0% (by GC) stabilised

供应商: TCI
Warning

Synonyms: Methacrylic Acid Hexyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M0503-25ML M0503-500ML
TCIAM0503-25MLEA 0 CNY
TCIAM0503-25ML TCIAM0503-500ML
甲基丙烯酸正己酯 ≥98.0% (by GC) stabilised
甲基丙烯酸正己酯
分子式: C₁₀H₁₈O₂
沸点: 88°C
闪点: 80°C
MDL编号: MFCD00015283
CAS号: 142-09-6
EINECS: 205-521-9

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR