Variamine blue B sulphate (=4-amino-4'-methoxydiphenylamine sulphate) ≥97.0% (by total nitrogen basis) for iron-titration

供应商: TCI

Synonyms: 4-Amino-4'-methoxydiphenylamine Sulphate, N-(4-Methoxyphenyl)-1,4-phenylenediamine sulphate

M0495-1G M0495-25G
TCIAM0495-1GEA 0 CNY
TCIAM0495-1G TCIAM0495-25G
Variamine blue B sulphate (=4-amino-4'-methoxydiphenylamine sulphate) ≥97.0% (by total nitrogen basis) for iron-titration
Variamine blue B sulphate (=4-amino-4'-methoxydiphenylamine sulphate)
分子式: C₁₃H₁₄N₂O
分子量: 312.34 g/mol
MDL编号: MFCD00060258
CAS号: 3169-21-9

现在订购

产品技术规格

Appearance Gray to Dark blue to Black powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Sensitiveness Colorimetric Test : Turn Blue purple with 1ppm Iron and turn Colorless to Almost colorless by adding EDTA 2Na


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR