Sodium-N-methyltaurinate 62 - 66% in water

供应商: TCI

Synonyms: Ethanesulphonic acid 2-(methylamino) sodium salt (1:1), Sodium 2-(methylamino)ethanesulphonate, N-Methyltaurine sodium salt

M0434-25G M0434-500G
TCIAM0434-25GEA 0 CNY
TCIAM0434-25G TCIAM0434-500G
Sodium-N-methyltaurinate 62 - 66% in water
Sodium-N-methyltaurinate
分子式: C₃H₈NNaO₃S
MDL编号: MFCD00066598
CAS号: 4316-74-9
EINECS: 224-339-0
Merck Index: 14,06124

现在订购

产品技术规格

Concentration (with Total Nitrogen) 62.0 to 66.0 w/w%
Free Alkali max. 17.0 % (as NaOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR