Poly(methyl methacrylate)

供应商: TCI

M0088-25G M0088-500G
TCIAM0088-25GEA 0 CNY
TCIAM0088-25G TCIAM0088-500G
Poly(methyl methacrylate)
Poly(methyl methacrylate)
分子式: C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
MDL编号: MFCD00134349
CAS号: 9011-14-7

现在订购

产品技术规格

Appearance White powder to crystal
Solubility in THF very faint turbidity
Viscosity 2.0 to 4.0 mPa-s (0.5g/50mL THF, 20 deg-C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR