内消旋酒石酸 ≥90.0% (by HPLC)

供应商: TCI
Warning

Synonyms: 中酒石酸

M0070-25G M0070-5G
TCIAM0070-25GEA 0 CNY
TCIAM0070-25G TCIAM0070-5G
内消旋酒石酸 ≥90.0% (by HPLC)
内消旋酒石酸
分子式: C₄H₆O₆
分子量: 150.09 g/mol
MDL编号: MFCD00063641
CAS号: 5990-63-6
EINECS: 205-696-1
Merck Index: 14,09071

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 90.0 area%
Water 8.0 to 11.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR