D(+)-Melibiose monohydrate ≥99.0% (by HPLC)
供应商: TCI
Synonyms:
6-O-α-D-Galactopyranosyl-D-glucopyranose monohydrate, (2R,3S,4R,5R)-2,3,4,5-Tetrahydroxy-6-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, alpha-D-(+)-Melibiose monohydrate
M0050-1G
M0050-10G
TCIAM0050-1GEA
0
CNY
TCIAM0050-1G
TCIAM0050-10G
D(+)-Melibiose monohydrate ≥99.0% (by HPLC)
D-(+)-Melibiose monohydrate
分子式:
C₁₂H₂₂O₁₁·H₂O |
MDL编号:
MFCD00198188 CAS号: 66009-10-7 EINECS: 209-568-6 Merck Index: 14,05820 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) (CAD) | min. 99.0 area% |
Specific rotation [a]20/D | +132 to +139 deg (C=4, H2O) |
Water | 4.0 to 6.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...