Lamivudine ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: (-)-1-[(2R,5S)-2-(Hydroxymethyl)-1,3-oxathiolan-5-yl]cytosine, 4-Amino-1-((2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one, 3TC

L0217-1G L0217-100MG
TCIAL0217-1GEA 0 CNY
TCIAL0217-1G TCIAL0217-100MG
Lamivudine ≥98.0% (by HPLC, titration analysis)
Lamivudine
分子式: C₈H₁₁N₃O₃S
存储条件: Freezer
MDL编号: MFCD00869739
CAS号: 134678-17-4
Merck Index: 14,05352

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C
Specific rotation [a]20/D -137.0 to -147.0 deg (C=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR