L-Leucine-tert-butyl ester hydrochloride ≥98.0% (by titrimetric analysis)
供应商: TCI
Synonyms:
(S)-tert-Butyl 2-amino-4-methylpentanoate hydrochloride, H-Leu-OtBu·HCl, L-leucine 1,1-dimethylethyl ester hydrochloride, H-Leu-OtBu.HCl, TERT-BUTYL (2S)-2-AMINO-4-METHYLPENTANOATE HYDROCHLORIDE
L0197-1G
L0197-5G
TCIAL0197-1GEA
0
CNY
TCIAL0197-1G
TCIAL0197-5G
L-Leucine-tert-butyl ester hydrochloride ≥98.0% (by titrimetric analysis)
L-Leucine-tert-butyl ester hydrochloride
分子式:
C₁₀H₂₁NO₂ 分子量: 223.74 g/mol 溶点: 167°C |
MDL编号:
MFCD00038906 CAS号: 2748-02-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Specific rotation [a]20/D | +18.0 to +21.0 deg (C=2, EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...