H-D-Leu-OtBu HCl ≥98.0% (by total nitrogen and titration analysis)

供应商: TCI

Synonyms: H-D-Leu-OtBu·HCl, (R)-tert-Butyl 2-amino-4-methylpentanoate hydrochloride

L0196-1G L0196-5G
TCIAL0196-1GEA 0 CNY
TCIAL0196-1G TCIAL0196-5G
H-D-Leu-OtBu HCl ≥98.0% (by total nitrogen and titration analysis)
H-D-Leu-OtBu HCl
分子式: C₁₀H₂₂ClNO₂
分子量: 223.7402 g/mol
MDL编号: MFCD00083642
CAS号: 13081-32-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D -15.0 to -23.0 deg (C=2, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR