L-tert-leucine methylamide ≥96.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
L-tert-Leucylaminomethane, (S)-2-Amino-N,3,3-trimethylbutanamide
L0168-1G
L0168-5G
TCIAL0168-1GEA
0
CNY
TCIAL0168-1G
TCIAL0168-5G
L-tert-leucine methylamide ≥96.0% (by titrimetric analysis)
L-tert-leucine methylamide
分子式:
C₇H₁₆N₂O |
MDL编号:
MFCD00673329 CAS号: 89226-12-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 96.0 % |
Specific rotation [a]20/D | +52.0 to +58.0 deg (C=0.5, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...