(S)-(-)-乳酸苄酯 ≥98.0% (by GC)

供应商: TCI

Synonyms: Benzyl L-lactate, (S)-(-)-Lactic Acid Benzyl Ester, Benzyl (S)-Lactate

L0161-25G L0161-5G
TCIAL0161-25GEA 0 CNY
TCIAL0161-25G TCIAL0161-5G
(S)-(-)-乳酸苄酯 ≥98.0% (by GC)
(S)-(-)-乳酸苄酯
分子式: C₁₀H₁₂O₃
分子量: 180.20 g/mol
沸点: 141°C
溶点: 17°C
MDL编号: MFCD00209546
CAS号: 56777-24-3

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -16.0 to -23.0 deg (C=1, Acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR