Loperamide hydrochloride ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Danger

L0154-25G L0154-5G
TCIAL0154-25GEA 0 CNY
TCIAL0154-25G TCIAL0154-5G
Loperamide hydrochloride ≥98.0% (by HPLC, titration analysis)
Loperamide hydrochloride
分子式: C₂₉H₃₃ClN₂O₂
分子量: 513.50 g/mol
溶点: 223°C
MDL编号: MFCD00058581
CAS号: 34552-83-5
EINECS: 252-082-4
UN号: 2811
ADR: 6.1 III
Merck Index: 14,05571

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR