L-Leucinamide hydrochloride ≥98.0% (通过滴定分析)

供应商: TCI

L0094-1G L0094-5G
TCIAL0094-1GEA 0 CNY
TCIAL0094-1G TCIAL0094-5G
L-Leucinamide hydrochloride ≥98.0% (通过滴定分析)
L-Leucinamide hydrochloride
分子式: C₆H₁₄N₂O
MDL编号: MFCD00013012
CAS号: 10466-61-2
EINECS: 233-952-2

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +8.5 to +11.0 deg (C=5, H2O)
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR