Laminaran (from Eisenia bicyclis)
供应商: TCI
L0088-1G
L0088-25G
TCIAL0088-1GEA
0
CNY
TCIAL0088-1G
TCIAL0088-25G
Laminaran (from Eisenia bicyclis)
Laminaran
分子式:
(C₆H₁₀O₅)n |
MDL编号:
MFCD00466918 CAS号: 9008-22-4 EINECS: 232-712-4 Merck Index: 14,05350 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Specific rotation [a]20/D | -40.0 to -50.0 deg (C=0.9, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...