久洛利定 ≥97.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
2,3,6,7-四氢-1H,5H-苯并[i,j]喹嗪
J0007-25G
J0007-5G
TCIAJ0007-25GEA
0
CNY
TCIAJ0007-25G
TCIAJ0007-5G
久洛利定 ≥97.0% (by GC, titration analysis)
久洛利定
分子式:
C₁₂H₁₅N 分子量: 173.26 g/mol 沸点: 280°C 存储条件: Refrigerator |
MDL编号:
MFCD00006917 CAS号: 479-59-4 EINECS: 207-535-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 35.0 to 39.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...