4-Isopropoxybenzenesulphonyl chloride ≥98.0% (通过GC)

供应商: TCI
Danger

I0802-25G I0802-5G
TCIAI0802-25GEA 0 CNY
TCIAI0802-25G TCIAI0802-5G
4-Isopropoxybenzenesulphonyl chloride ≥98.0% (通过GC)
4-Isopropoxybenzenesulphonyl chloride
分子式: C₉H₁₁ClO₃S
沸点: 113°C
存储条件: Refrigerator
MDL编号: MFCD03093083
CAS号: 98995-40-5
UN号: 3261
ADR: 8,II

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 95.0 %
Melting point 36.0 to 40.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR