L-Isoleucinamide hydrochloride ≥98.0% (以总氮计)

供应商: TCI

I0731-1G I0731-5G
TCIAI0731-1GEA 0 CNY
TCIAI0731-1G TCIAI0731-5G
L-Isoleucinamide hydrochloride ≥98.0% (以总氮计)
L-Isoleucinamide hydrochloride
分子式: C₆H₁₄N₂O
MDL编号: MFCD00058476
CAS号: 10466-56-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +19.0 to +25.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR