1,2-O-异亚丙基-α-D-呋喃木糖 ≥98.0% (by GC)

供应商: TCI

Synonyms: 1,2-O-Isopropylidene-α-D-xylofuranose, (3aR,5R,6S,6aR)-5-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-ol, (3aR,5R,6S,6aR)-5-(Hydroxymethyl)-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-6-ol

I0721-25G I0721-5G
TCIAI0721-25GEA 0 CNY
TCIAI0721-25G TCIAI0721-5G
1,2-O-异亚丙基-α-D-呋喃木糖 ≥98.0% (by GC)
1,2-O-异亚丙基-α-D-呋喃木糖
分子式: C₈H₁₄O₅
MDL编号: MFCD00063295
CAS号: 20031-21-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C
Specific rotation [a]20/D -16.0 to -22.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR