L-Isoleucine-tert-butyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
供应商: TCI
Synonyms:
H-Ile-OtBu.HCl, tert-Butyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
I0685-1G
I0685-5G
TCIAI0685-1GEA
0
CNY
TCIAI0685-1G
TCIAI0685-5G
L-Isoleucine-tert-butyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
L-Isoleucine-tert-butyl ester hydrochloride
分子式:
C₁₀H₂₁NO₂ 分子量: 223.74 g/mol 溶点: 160°C |
MDL编号:
MFCD00058015 CAS号: 69320-89-4 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Specific rotation [a]20/D | +13.0 to +17.0 deg (C=1, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...