2-(1H-Imidazol-1-yl)acetic acid ≥98.0% (通过GC,滴定分析)

供应商: TCI

I0670-25G I0670-5G
TCIAI0670-25GEA 0 CNY
TCIAI0670-25G TCIAI0670-5G
2-(1H-Imidazol-1-yl)acetic acid ≥98.0% (通过GC,滴定分析)
2-(1H-Imidazol-1-yl)acetic acid
分子式: C₅H₆N₂O₂
溶点: 269°C
MDL编号: MFCD01721800
CAS号: 22884-10-2

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR